Would you like to log in again?
Compact Glass Slide Storage Box; The Mini Box, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71455-B 71455-W 71455-R
100501-428EA 4.75 USD InStock
100501-428 100501-426 100501-424
Compact Glass Slide Storage Box; The Mini Box, Electron Microscopy Sciences
Microscope Slide Boxes
Slide box made from rigid polystyrene, with a removable lid.
This 25 capacity slide box measures L x W x H: 95 x 82 x 30 mm (7.75 x 3.25 x 1.25"). The box is available in either white, red, or blue.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty