In Copper (Cu), Nickel (Ni), Gold (Au), Copper/Palladium (Cu/Pd=CP), Molybdenum (Mo).
- Measures 3.05 mm in diameter
- 0.7 mil (18 µm) thickness
G200TH is a thickened version of the standard, with an asymmetric centre. A mark on the rim allows for precise orientation of the grids.
G200TT is a combination of thin and thick bar grids, with a mark on the rim for orientation.
G200T is designed for ease of handling and is easily removed if necessary. To remove the handle, just bend on a 90 degree angle.