Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sample Storage Sets; Poly-Seal Screw Cap, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64270 64271
100496-178EA 208.7 USD InStock
100496-178 100501-584
Sample Storage Sets; Poly-Seal Screw Cap, Electron Microscopy Sciences
Vials
Threaded glass vials with Poly-seal caps and a cone shaped polyethylene liner.
Alpha-numerical indexing system allows quick identification and location of stored items.
Delivery information: Each set includes a partitioned box, vials with caps, labels, reference chart, and an exterior card holder and card.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty