Would you like to log in again?
Limonene-Mount, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
17987-01 17987-06 17987-25
100501-606EA 1325.9 USD InStock
100501-606 100496-554 100496-556
Limonene-Mount, Electron Microscopy Science
Stains Microscopy and staining reagents
This mounting medium is made with limonene, a natural product from orange peels. It preserves  tissues and cell smears that can be dehydrated with organic solvents in immunohistochemistry, e.g. DAB and DAB with nickel or cobalt.
  • A direct substitute for BioMeda™ Clarion Mount
  • Use for mounting tissues and cell smears
  • Works well with alkaline phosphatase chromogens, and organic solvent resistant Supper Fast Red
  • A good choice for mounting H & E stained slides.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty