Would you like to log in again?
Miniature Measuring Scales, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62135-03 62135-01 62135-02
100503-066EA 76.17 USD InStock
100503-066 100501-686 100497-900
Miniature Measuring Scales, Electron Microscopy Sciences
Spatulas
These unit measuring scales may be used directly under a microscope or magnifier for quick measuring of small specimens, parts, grooves, slots, line widths, holes, chambers, and more.
Precision fabricated from stainless steel, the scales are welded to bendable stainless steel shafts and mounted in color coded, non-rolling handles.
Packaging: Each scale is individually packed in a plastic tube.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty