Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Sample Vials, Clear And Amber, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60993-08 60993-01 60992-04 60992-16 60992-24 60992-01 60993-04 60992-02 60992-08 60992-12 60993-02
100500-276CS 540.78 USD InStock
100500-276 100500-266 100500-278 100501-738 100500-268 100503-122 100500-270 100500-272 100500-262 100500-274 100500-264
Sample Vials, Clear And Amber, Electron Microscopy Sciences
Vials
Multi purpose glass sample vials suited for research studies. Clear vials are manufactured from Wheaton-33R a low extractable borosilicate glass which protects against changes in the pH and maintains sample integrity. Amber vials are manufactured from Wheaton '320' borosilicate glass and protects light-sensitive samples.
  • Economically priced
  • Come with lined caps
  • Can be sterilized
  • Can be stored at low temperatures
Packaging: Packaged in two shrink-wrapped packs of 100 vials. Caps are packaged separately.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty