Would you like to log in again?
Centrifugal Filter Devices - Costar Brand , Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67030-61 67031-70 67030-63 67030-60 67031-69 67030-62
100500-510CS 506.2 USD InStock
100500-510 100500-512 100500-514 100500-516 100501-762 100501-760
Centrifugal Filter Devices - Costar Brand , Electron Microscopy Sciences
Filters Centrifugal Filters
Spin-X centrifuge tube filters utilize centrifugation for the filtering of small volumes.
  • Filter samples in microcentrifuges
  • Choice of cellulose or nylon membrane
  • For volume up to 500 µl
The unit consists of a polypropylene filter insert and a centrifuge tube. They have a cellulose acetate membrane for aqueous solutions which is ideal for the removal of bacteria, particles, and cells from media. They are also effective for DNA removal from agarose or acrylamide gels. Spin-X LC units have a nylon membrane for applications such as HPLC sample preparation. Both have retailed volume of less than 5 µl, and withstand relative centrifugal force (RFC) to 16000×g.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty