Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Due to scheduled system maintenance, vwr.com will be temporarily unavailable from 6:00 am EST to 2:00 PM EST on Saturday, May 25. 

Magnetic Stirring Bars, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences
64510-16 64510-13 64510-24 64510-05 64510-22 64510-12 64510-18
100501-010BX 140.01 USD InStock
100501-010 100501-778 100501-008 100501-016 100501-014 100503-156 100501-012
Magnetic Stirring Bars, Electron Microscopy Sciences
Stirrers Magnetic Stirring Bars
Round Assortment Box: 22 round magnetic stirring bars

PTFE with removable pivot ring on 1" long and over. The box consists of 2 each: 1/2 x 3/4", 1 x 5/16", 2 x 3/8", 1/2 x 5/16", 3/4 x 5/16", 1 x 3/8", 1 1/2 x 5/16", 1 3/4 x 5/16", 2 x 5/16" and 1 each: 1/4 x 5/16", 1/4 x 3/8", 1 1/2 x 3/8", 1 3/4 x 3/8". Octagon Assortment Box with Micro Spinbars: Box contains 16 assorted bars; It consists of 12 octagon magnetic stirring bars, "PTFE" with pivot ring and 4 fleas. 2 each: 7 x 2 mm, 10 x 3 mm, 1/2 x 5/16", 5/8 x 5/16", 1 x 3/8", 1 1/2 x 3/8", 2 x 3/8", 2 1/2 x 3/8".

Round Assortment Box: 22 round magnetic stirring bars. "PTFE" with removable pivot ring on 1" long and over. The box consists of 2 each: 1/2 x 3/4", 1 x 5/16", 2 x 3/8", 1/2 x 5/16", 3/4 x 5/16", 1 x 3/8", 1 1/2 x 5/16", 1 3/4 x 5/16", 2 x 5/16" and 1 each: 1/4 x 5/16", 1/4 x 3/8", 1 1/2 x 3/8", 1 3/4 x 3/8".

Metric Polygon Assortment Box: Polygon stirring bars: "PTFE". Consists of 2 each: 10 x 6 mm, 15 x 6 mm, 20 x7 mm, 25 x 8 mm, 30 x 8 mm, 40 x 8 mm, 50 x 8 mm, 60 x 8 mm, 80 x 10 mm.

Color Coded Octagon Assortment Box: 24 color coded octagon magnetic stirring bars: "PTFE". with pivot ring. Consists of 1 each: Yellow, Red And Blue: 1/2 x 1/8”, 5/8 x 5/16”, 7/8 x 5/16”, 1/2 x 5/16”, 1 x 5/16”, 1 1/2 x 5/16”, 2 x 5/16”, 3 x 1/2”.

Egg-Shaped Assortment Box: These egg-shaped magnetic stirring bars provide its own pivot point. Designed especially for use in round-bottom vessels. 13 Egg-shaped spin bars, "PTFE". Consists 1 each: 3 x 3/4", 2 each: 3/4 x 3/8", 1 x 1/2", 1/4 x 5/8", 1 1/2 x 5/8", 1 5/8 x 3/4", 2 x 3/4".

SpinPlus ™ Assortment Box: Add speed and efficiency to your mixing. The "+" shape creates a deep vortex and provides stability to your mixing. 5 SpinPlus magnetic stirring bars, "PTFE". Consists of 1 each: 3/8 x3/8", 3/4 x 3/4", 1 x 1",1 1/4 x 1 1/4", 1 1/2 x 1 1/2".

Colored Micro Assortment Box: 12 Micro Spin Bar magnetic stirring bars in assorted colors; "PTFE". Consists of 1 each of every color and size: red, yellow and blue: 8 x 1.5 mm, 5 x 2mm, 7 x 2 mm, 10 x 3 mm. Metric Polygon Assortment Box: Polygon stirring bars: "PTFE". Consists of 2 each: 10 x 6 mm, 15 x 6 mm, 20 x7 mm, 25 x 8 mm, 30 x 8 mm, 40 x 8 mm, 50 x 8 mm, 60 x 8 mm, 80 x 10 mm.

Color Coded Octagon Assortment Box: 24 color coded octagon magnetic stirring bars: "PTFE". with pivot ring. Consists of 1 each: Yellow, Red And Blue: 1/2 x 1/8”, 5/8 x 5/16”, 7/8 x 5/16”, 1/2 x 5/16”, 1 x 5/16”, 1 1/2 x 5/16”, 2 x 5/16”, 3 x 1/2”.

Egg-Shaped Assortment Box: These egg-shaped magnetic stirring bars provide its own pivot point. Designed especially for use in round-bottom vessels. 13 Egg-shaped spin bars, "PTFE". Consists 1 each: 3 x 3/4", 2 each: 3/4 x 3/8", 1 x 1/2", 1/4 x 5/8", 1 1/2 x 5/8", 1 5/8 x 3/4", 2 x 3/4".

SpinPlus ™ Assortment Box: Add speed and efficiency to your mixing. The "+" shape creates a deep vortex and provides stability to your mixing. 5 SpinPlus magnetic stirring bars, "PTFE". Consists of 1 each: 3/8 x3/8", 3/4 x 3/4", 1 x 1",1 1/4 x 1 1/4", 1 1/2 x 1 1/2".

Colored Micro Assortment Box: 12 Micro Spin Bar magnetic stirring bars in assorted colors; "PTFE". Consists of 1 each of every color and size: red, yellow and blue: 8 x 1.5 mm, 5 x 2mm, 7 x 2 mm, 10 x 3 mm.

Order Now
 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty