Would you like to log in again?
Wide Mouth Cryogenic Vial, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
61850-15
100501-980PK 326.85 USD InStock
100501-980
Wide Mouth Cryogenic Vial, Electron Microscopy Sciences
Vials
These wide mouth cryogenic vials are useful for the storage and retrieval of solid specimens, such as bone, tumor, and organ samples.
  • Selfstanding
  • Radiation sterilized, noncytotoxic, non-pyrogenic
Made from polypropylene with high- density polyethylene screw closures.
Ordering information: Caps are attached in 15 bags of 5 vials each or 75 per case.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty