Would you like to log in again?
Biopsy Bags, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62326-10 62326-05 62326-03 62326-01 62326-02 62326-15
103302-090PK 82.55 USD InStock
103302-090 100490-170 100501-992 103302-474 103302-550 103301-906
Biopsy Bags, Electron Microscopy Sciences
Bags Sample Bags
Bags are made up of porous paper which provides maximum tissue safety during processing.
  • Made from porous paper
  • Permits proper fluid exchange and drainage
  • Available in 25 and 35 mm diameter or 25 and 32 mm sizes
They permit proper fluid exchange and drainage.
To use: simply introduce the specimen into the bag and fold it enclosing the cassette or capsule. Now ready to process as usual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty