Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Disposable Sterile Vacuum Filter Units, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
67047-12 67046-15 67048-13 67045-11 67046-11 67046-14 67045-13 67048-14 67045-12 67048-16 67046-13 67047-11 67047-14 67048-12 67046-12 67047-15 67048-15 67047-13
100490-682PK 296.99 USD InStock
100490-682 100490-692 100490-684 100490-672 100490-694 100490-680 100490-690 100502-026 100490-704 100490-678 100490-700 100490-702 100490-686 100490-674 100490-696 100490-688 100490-698 100490-676
Disposable Sterile Vacuum Filter Units, Electron Microscopy Sciences
Filters Bottle Top Filters
Ideal for filtration of tissue culture media, biological fluids, fixation buffers and more. (not for parenteral use.)
  • Vacuum filtration units fit standard threaded bottle tops
  • Designed for use on glass or polystyrene media bottles
  • Attaches to most popular 33 and 45 mm Ø bottle's necks with standard GPI threads
Units are color coded by membrane type (CA: Orange; NYL: Red, CN: Blue; and PES: Yellow). Choice of membrane media: Cellulose acetate (CA), for low protein binding. Nylon (NYL), for low extractable. Cellulose Nitrate (CN), for general purpose filtration. Or polyethersulfone (PES), for fast flow rates and low protein binding.
Membranes come in 0.20, 0.22 or 0.45 µm pore sizes. All four-type membranes are detergent-free, tissue culture compatible, and heat-sealed to the support grid to maximize flow rate, reduce foaming and protein denaturization.
Ordering information: The unit includes membrane filter, glass fiber pre-filter and graduated funnel of clear polystyrene with polyethylene neck adapter. Systems are individually packed, gamma irradiation sterilized and certified non-pyrogenic. Comes with tubing adapter that fits most vacuum hoses.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty