Would you like to log in again?
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70310 70310-02 70310-06
100492-918EA 700.64 USD InStock
100492-918 102096-604 100502-078
Desk Tally Counter, Four Digit, Electron Microscopy Sciences
Cash and Check Handling Supplies Tally Counters
The desk tally counter comes with one, two, or six counting units.
One Counting Unit: A chrome plated with counter with a flange (21/2" dia.) for desktop mounting. The recording lever is on the right side and the reset knob on the top. The counter is ideal for serial sectioning work. Four digit place.
Two Counting Units: A Counter with two counting keys each with 0 to 999 range with a single reset knob it comes with removable tabs for top identification.
Six Counting Units: A counter with five keys and one totalize unit (0 to 999), which records the total number of strokes made on other keys. At 100 increments, totalizing unit rings bell. When totalizing unit records 100, each figure corresponding to individual keys automatically displays percentage. Two reset knobs, removable tabs on top of counter provide area for identification of clasification being counted.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty