Would you like to log in again?
Ultrastructure Size Calculator, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
74804
100502-130EA 10.69 USD InStock
100502-130
Ultrastructure Size Calculator, Electron Microscopy Sciences
Calculators Pocket Calculators
A slide-guide-calculator used to determine the real-life size of a structure from a micrograph- in micrometer, nanometer or angstrom units
It computes the real-life dimension/mm at magnifications from 1,000 to 100,000 by introducing a multiplication factor of x2 or x.5. The magnification range can be extended. Waterproof card, printed in English, German and French.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty