Would you like to log in again?
Dumont Diamond Tips Straight Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78100
100502-140EA 80.73 USD InStock
100502-140
Dumont Diamond Tips Straight Tweezers, Electron Microscopy Sciences
Forceps
Dumont Stainless Steel tweezers coated with an extremely fine diamond dust in the inside tip.
Ideal for applications where a tight grip is necessary. They are also available with an improved locking system. May not be suitable for use with EM grids.
Fine Tip: Tip measures 0.25 by 0.25 mm.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty