Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
EMS Surface Mount and Optoelectric Tweezers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78250-116 78250-110 78250-60 78250-104 78250-117 78250-106 78250-100 78250-90 78250-70 78250-2 78250-50 78250-10 78250-40 78250-109 78250-K 78250-3
100502-292EA 43.41 USD InStock
100502-292 76376-134 100502-294 76376-136 100502-284 76376-128 76376-138 100502-290 76376-140 100502-296 76376-142 76376-130 100502-286 100502-298 100502-288 76376-132
EMS Surface Mount and Optoelectric Tweezers, Electron Microscopy Sciences
Forceps
These tweezers are made from SA low carbon austenitic steel that is non magnetic and anti acid which is ideal for use where corrosion resistance and toughness are primary requirements.
They offer comfort and smooth handling.They are available in different tips and angles for specific jobs and they have a satin antiglare finish with an ergonomic design.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty