Would you like to log in again?
Apiezon Grease, Type AP101, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60706-40 60706-01
100497-460EA 4241.23 USD InStock
100497-460 100502-458
Apiezon Grease, Type AP101, Electron Microscopy Sciences
Lubricants
Type 101 is an excellent general purpose, hydrocarbon grease, which is intended for a variety of industrial and scientific applications.
Non silicone Grease: Being hydrocarbon based AP 101 does not suffer from the problems of "creep" or "carry over" which is traditionally associated with silicone greases.
Anti-Seize Properties: The AP 101 contains "PTFE" which confers superior anti-seize properties providing long lasting lubrication and ensuring smooth operation of stop cocks and taps. The value of AP 101 anti-seize properties are not limited to laborat.
Non silicone Grease: Being hydrocarbon based AP 101 does not suffer from the problems of "creep" or "carry over" which is traditionally associated with silicone greases.
Solvent resistant: Both "PTFE" and the lithium stearate gel base in AP 101 are insoluble in most solvents. They ensure that AP 101 shows resistance to water, alcohols, ketones and esters.
Wide temperature range: It can be used over a very wide range of temperatures, possessing its optimum consistency over the -15 to +150°C temperature range, but is usable down to -40°C and, for limited periods, up to +180°C.
Under vacuum: AP 101 exhibits good vacuum properties, down to 10-4 torr.
Easily remove AP 101 by wiping it with a soft cloth. Any residues of grease can be washed away with an aqueous glassware detergent.
Solvent resistant: Both "PTFE" and the lithium stearate gel base in AP 101 are insoluble in most solvents. They ensure that AP 101 shows resistance to water, alcohols, ketones and esters.
Wide temperature range: It can be used over a very wide range of temperatures, possessing its optimum consistency over the -15 to +150°C temperature range, but is usable down to -40°C and, for limited periods, up to +180°C.
Easily remove AP 101 by wiping it with a soft cloth. Any residues of grease can be washed away with an aqueous glassware detergent.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty