Would you like to log in again?
Mounting Media/Canada Balsam Bottles, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64211-01 64211-06
100502-536EA 92.88 USD InStock
100502-536 100490-418
Mounting Media/Canada Balsam Bottles, Electron Microscopy Sciences
Bottles Cell Culture Bottles
Ideal for applying mounting media. Cap is ground to the shoulder of the bottle to form a seal.
  • Made from soda lime glass
  • 100 ml capacity
Delivery information: The bottle comes with a glass applicator rod and glass cap.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty