Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
MeshWell Plates, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64703-01 64700-24 64700-12 64702-06 64700-06 64702-12 64702-24
103306-230EA 1694.59 USD InStock
103306-230 102095-518 100502-548 100498-850 102095-514 102095-516 100498-848
MeshWell Plates, Electron Microscopy Sciences
Microplates Cell Culture Plates
These plates are made from polypropylene and the bottom of each of the wells is an ETFE monofilament screening fabric with a 500 µm opening.
  • Available with 6, 12, and 24 well configurations
  • Also able to process tissue slices with Reagent Trays
Fits into the 6, 12 and 24 Costar/Corning Cell Culture Clusters for processing tissue slices.
Ordering information: The MeshWell Plate set includes one MeshWell and one Cluster plate.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty