Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formalin 10%, TissuePrep, buffered

Supplier: Electron Microscopy Sciences
Danger
Synonyms: MethylaldehydeFormolParaformaldehyde aqueous solutionFormaldehyde

15742-30 15742-60 15742-10 15742-20
100500-248CS 98.58 USD InStock
100500-248 100502-586 100502-588 100503-120
Formalin 10%, TissuePrep, buffered
Formalin
Ready-to-use, prefilled with 10% formalin.

Prefilled with 10% formalin Cat# 15740. Ready-to-use in pre-filled cups. Labeled in accordance to OSHA requirements, which gives you immediate information regarding handling and first aid procedures. Pre-measured in wide-mouth, leak-proof, opaque polypropylene containers which limits your exposure to formaldehyde and saves you time.


Formula: CH₂O
MW: 30.03 g/mol
MDL Number: MFCD00003274
CAS Number: 50-00-0
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty