Would you like to log in again?
Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.
SecureSlip™ Cell Culture Coverglass, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
70462-14 70462-12
100502-726PK 673.06 USD InStock
100502-726 102681-198
SecureSlip™ Cell Culture Coverglass, Electron Microscopy Sciences
Cover Glasses
Convenient, Secure coverglass that will not move during cultivation
The coverglass is affixed to a thin microscopically transparent silicone base which secure to culture vessels by an electrostatic charge, not an adhesive, to prevent movement during cultivation. The silicone also acts as a hydrophobic barrier to isolate liquid reagents on glass. Individual coverglass with silicone backing can be adhered to a petri dish or plate. These can be easily removed one at a time from the dish during culture. Once removed, coverglass can be affixed to a microscope slide by means of the tacky silicone backing for routine processing, or placed cell side down onto a drop of reagent using the silicone backing as a cover to prevent evaporation.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty