A thin film of pure carbon is deposited on the shiny side of the grid.
- Select copper, nickel, or gold grids in 150, 200, 300, or 400 mesh
- Optically checked followed by batch testing in the EM
Carbon coatings thickness of Standard: Approx. 5-6nm, Ultra-Thin (UL): 3-4nm, Thick (TH): 10nm, Extra Thick (ET): 20-30nm.
Ordering information: Available in Standard (5 - 6 nm), Ultra-Thin (3 - 4 nm), Thick (10 nm) and Extra Thick (20 - 30 nm) film thickness.
Packaging: Packed in grid storage box.