Would you like to log in again?
Pap-Pen; Slide Marker, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
71310 71312
100502-806EA 100.4 USD InStock
100502-806 100491-772
Pap-Pen; Slide Marker, Electron Microscopy Sciences
Stains Microscopy and staining reagents
New and improved staining guard pen.
New and improved staining guard pen. The Super PAP Pen outperforms the original PAP Pen. The Super PAP Pen may be used at temperatures as high as 120°F. (The old style PAP Pen worked only up to 90°F). With this high temperature feature, one can now use the Super PAP Pen to perform the peroxidase-anti-peroxidase method, ABC method, and Fluorescent antibodies method with satisfactory results.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty