Would you like to log in again?
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
70175-40 70175-50 70175-60
101454-078PK 614.33 USD InStock
101454-078 100502-820 101454-080
Polyethylene Molds, Embedding Stubs, Electron Microscopy Sciences
Moulds Moulds for Histology
Block holders molded from hard plastic are easy to store as it attched to Aluminum chucks (ABH)
The large center hole in the holder prevents bubble formation and its frosted surface is easy for marking.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty