Would you like to log in again?
Russian Tissue Forceps, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
78260-06 78260-08
100494-660EA 64.21 USD InStock
100494-660 100502-880
Russian Tissue Forceps, Electron Microscopy Sciences
Forceps
Stainless steel, 155 mm (6”) long. With special cupped pattern for superior grip. Cup area is 6.5 mm wide and 9 mm long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty