Gilder grids are a reliable support specimen grid for transmission electron microscopy. They feature a well-defined grid bars, maximum open area, and a matt or shiny side. Each grid is individually inspected.
- 3.05 mm diameter
- 0.7 mil (18 µm) thickness
- In Copper (Cu), Nickel (Ni), or Gold (Au)