Would you like to log in again?
Micro Spoon Heyman Type, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
62411-B 62411-A
100503-080EA 7.77 USD InStock
100503-080 100503-078
Micro Spoon Heyman Type, Electron Microscopy Sciences
Spatulas
A Heyman type micro spatula made from nickel stainless steel with a glazed finish.
  • Wire Ø - 2.62 mm (0.103")
  • Overall length: 159 mm (6¹/₄")
  • Flat end: 31.35 mm (1¹/₄") long
The spoon end: 12.5 mm (½") long × 3 mm (¹/₈") wide. Type b has a sharpened bent end: 37.5 mm (1½") long. V-Shaped formed other end: ½" long.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty