Would you like to log in again?
Sharp Objects Disposal-Pouch, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64430 64432 64431
100496-212EA 49.38 USD InStock
100496-212 100503-172 100503-170
Sharp Objects Disposal-Pouch, Electron Microscopy Sciences
Bags Waste Disposal Bags
Made of 10 mil solid paperboard, which opens to an oval shape by folding the bottom flaps.
  • Ideal for needles, razor blades, scalpel blades, more
When full just close the top flaps and lock into place. An epoxy coated steel wire pouch stand provides stable support to that keeps the pouch standing upright.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty