Would you like to log in again?
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
24145-25 24145-10
100503-194EA 41.92 USD InStock
100503-194 100503-188
RED-Z - Fluid Control Solidifier, Electron Microscopy Sciences
Spill Control Materials
RED-Z Solidifier will solidify, decontaminate and deodorize any water base fluid at a ratio of ¾ oz to 1 L
It contains chlorine (NaDCC) an EPA registered disinfectant - almost twice the amount recommended by the CDC. Solidifier could also be used for blood and other body fluids. RED-Z available in a pointed top bottle that fits easily in porthole containers and in a flip-top shaker top bottle for large spills.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty