Would you like to log in again?
Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60950 60962 60960 60952
100503-198PK 132.1 USD InStock
100503-198 100503-196 100503-200 100503-278
Beakers Staccup; Plastic, Disposable, Electron Microscopy Sciences
Beakers
Made from heavy gauge polypropylene
  • Resistant to most chemicals
  • Frosted marking area
  • Customary and metric graduations
  • Anti-drip ridge
Suitable for parafilm cover.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty