Would you like to log in again?
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
64167 64151 64162 64157 64165 64161 64155 64153 64154
100503-314EA 18.42 USD InStock
100503-314 100503-304 102091-094 100503-224 100503-312 100503-268 100503-302 100503-310 100503-300
Wash Bottles, Permanently Labeled/Color-Coded, Electron Microscopy Sciences
Bottles Wash Bottles
Permanently labeled/color-coded wash bottles.
Pack of six - Variety includes: Acetone, Methanol, Water, Iso-propyl Alcohol, Ethanol and Toluene. Toluene Wash Bottle is for short term storage only.
These safe wash bottles, which are permanently labeled and color-coded, eliminate the risk of dispensing the wrong liquid by mistake. All of these wide-mouth (easy transfer liquid) polyethylene bottles have polyalomer caps and a fine-tip-nozzle that may be cut back for heavier flow. Capacity is 16 oz. (500 ml), low density polyethylene.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty