Would you like to log in again?
Glass Microbeakers, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
60986 60982 60984 60980
100503-380BX 657.12 USD InStock
100503-380 100503-374 100503-378 100503-376
Glass Microbeakers, Electron Microscopy Sciences
Beakers
Used for the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty