Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Formaldehyde 35.5 -38.0% stabilized ACS

Supplier: Electron Microscopy Sciences
Danger
Synonyms: MethylaldehydeFormolFormalinParaformaldehyde aqueous solution

Stabilization: Stabilized with methanol

15681 15682 15680
100503-910EA 122.11 USD InStock
100503-910 100503-402 100503-908
Formaldehyde 35.5 -38.0% stabilized ACS
Formaldehyde
  • Specific Gravity: 1.09


Formula: CH₂O
MW: 30.03 g/mol
Boiling Pt: 64 °C (147 °F)
Density: 1.09 g/ml
Flash Pt: 60 °C (140 °F
Storage Temperature: Ambient
MDL Number: MFCD00003274
CAS Number: 50-00-0
UN: 1198
ADR: 3,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/24/28984824.jpg
Order Now
Specification Test Results
Assay 35.5 to 38.0%
Residue after ignition 0.005%
Color (APHA) 10
Sulfate 0.002%
Heavy Metal (as Pb) 5ppm
Titratable acid 0.006 meq/g
Chloride (Cl) 5ppm
Iron (Fe) 5ppm

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty