Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Epo-Fix, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

1232 1245
100503-612EA 184.23 USD InStock
100503-612 100503-408
Epo-Fix, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Until now, LR White has been the most commonly used embedding media for materials due to the block hardness it offered, but there are difficulties associated with LR White

Formulated for material science. Epo-Fix is an old product that has existed only in the metallographic field until now. After extensive testing in our labs we have found that it is a perfect embedding media for a majority of material specimens.

Until now, LR White has been the most commonly used embedding media for materials due to the block hardness it offered, but there are difficulties associated with LR White. For instance, the adherance to the specimen is poor and additives are required to improve its adherance. Another major problem is the inability to orient the specimen correctly in order to cut you block afterwards. This is due to the fact that LR White needs to be oxygen starved and gelatin capsules (vertical) are recommended by the manufacturer for use. Another problem is that polymerization time in the oven can take over 24 hours. Epo-Fix remedies all of these problems. It has very good sectioning properties, good adherance to the sample, very low viscosity, easy curing in molds (no orientation problems), and quick cure times.

Epo-Fix Technical Data:
A two component epoxy resin, characterized by negligible shrinkage during curing and good mechanical properties in its hardened state. It is particularly suited for embedding hard samples of complicated shape and for vacuum impregnation. Samples such as paper, silicon, ceramics, metals, wafers, and chips are suitable to be used with Epo-Fix. It has low viscosity, low vapour pressure, good wetting properties, and minimal shrinkage. It hardens at room temperature without significant generation of heat in approximately 8 hours (no damage from heat or pressure), and at increased temperatures, 60°C, the hardening time is reduced to 2 hours. It should be noted that if you place Epo-Fix in a desiccator during curing you will eliminate any air bubbles.

Epo-Fix kit includes 1 kg resin, 120 gm hardener, 15 cups, 15 stirrers, 1 x 5 mL and 20 mL syringes.

Epo-Fix Technical Data:
A two component epoxy resin, characterized by negligible shrinkage during curing and good mechanical properties in its hardened state. It is particularly suited for embedding hard samples of complicated shape and for vacuum impregnation. Samples such as paper, silicon, ceramics, metals, wafers, and chips are suitable to be used with Epo-Fix. It has low viscosity, low vapour pressure, good wetting properties, and minimal shrinkage. It hardens at room temperature without significant generation of heat in approximately 8 hours (no damage from heat or pressure), and at increased temperatures, 60°C, the hardening time is reduced to 2 hours. It should be noted that if you place Epo-Fix in a desiccator during curing you will eliminate any air bubbles.

Epo-Fix kit includes 1 kg resin, 120 gm hardener, 15 cups, 15 stirrers, 1 x 5 mL and 20 mL syringes.

For instance, the adherance to the specimen is poor and additives are required to improve its adherance. Another major problem is the inability to orient the specimen correctly in order to cut you block afterwards. This is due to the fact that LR White needs to be oxygen starved and gelatin capsules (vertical) are recommended by the manufacturer for use. Another problem is that polymerization time in the oven can take over 24 hours. Epo-Fix remedies all of these problems. It has very good sectioning properties, good adherance to the sample, very low viscosity, easy curing in molds (no orientation problems), and quick cure times.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty