Would you like to log in again?
Araldite 6005, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Araldite®
13920
100503-424KT 88.23 USD InStock
100503-424
Araldite 6005, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Also known as American Araldite.
  • Araldite 6005 Kit
  • Blocks are slightly harder than those produced by Araldite 502
Kit consists of: 450 mL Araldite 6005, 450 mL DDSA; Specially Distilled, 100 mL BDMA, and 225 mL DBP.
*Please note that wherever DMP-30 is listed as the accelerator in our embedding media kits you have the option to use BDMA in place of it. BDMA is less viscous, has a longer shelf life, and offers better penetration (it should be noted, however, that in order to achieve the optimum results when using BDMA in place of DMP-30 you must vary the amount. DMP-30: 1.5-2%; BDMA: 2.5-3%).
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty