Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
HPMA, Water Soluble, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14220
100503-442KT 82.44 USD InStock
100503-442
HPMA, Water Soluble, Electron Microscopy Sciences
Stains Microscopy and staining reagents
HPMA is a water-soluble resin used for cytochemical applications
Kit consists of: 450 mL HPMA and 15g Benzoyl Peroxide Paste. Infiltration follows fixation of the tissue so there is no extraction of materials caused by dehydration in alcohols.
Leduc, E.H. et al., (1965) J. Cell Biol., 26:137.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty