Would you like to log in again?
Lowicryl Resin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
14350 14330 14360 14340
100503-448KT 1176.94 USD InStock
100503-448 100503-446 100503-452 100503-450
Lowicryl Resin, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Lowicryl resins are highly cross-linked acrylate-based embedding resins, designed for use over a wide range of embedding conditions. These resins provide low viscosity at low temperatures.
  • K4M kit is usable to –35 °C and is a polar (hydrophilic) medium
  • HM20 kit can be used down to –70 °C and is nonpolar (hydrophobic)
  • K11M has properties similar to K4M but can be at –60 °C
  • HM23 has properties similar to HM20 but can be used to at least –80 °C
All of these resins are photopolymerized by long wavelength (360 nm) ultraviolet light. Since the initiation of the polymerization is largely independent of temperatures, blocks may be polymerized at the same temperatures used for infiltration. The resin may also be chemically polymerized at 60 °C.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty