Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Histocryl Resin Kit, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

14390
100503-458KT 359.55 USD InStock
100503-458
Histocryl Resin Kit, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Histocryl acrylic resin has been specially formulated for light microscopy. It is water clear, and because it is hydrophilic (polar) it permits the use of most routine staining techniques without priorremoval or etching.

Kit consists of: 500 ml Histocryl Resin , 50g Histocryl Catalyst (B.P.) , 10 ml Histocryl Accelerator

Order Now
 

ORDER

SPECIFICATIONS

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty