Would you like to log in again?
Methyl Methacrylate/Butyl Methacrylate, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14520
100503-460KT 74.3 USD InStock
100503-460
Methyl Methacrylate/Butyl Methacrylate, Electron Microscopy Sciences
Stains Microscopy and staining reagents
The mixture of Butyl and Methyl Methacrylate offers many advantages over the standard GMA and HPMA embedding mediums
Kit consist of 225 mL Methyl Methacrylate, 225 mL Butyl Methacrylate, and 15 gm Benzoyl Peroxide Paste.
It can be UV cured in the cold with the addition of benzoin methyl ether, or heat cured by the addition of benzoyl peroxide. The resin can be removed from the sections with a solvent (acetone) before staining.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty