Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Quetol 651 - NSA, Electron Microscopy Sciences

Supplier: Electron Microscopy Sciences

REVIEWS (0)

Total Ratings: 0
Avg. Ratings: out of 5

Sort all by:

14640
100503-462KT 287.34 USD InStock
100503-462
Quetol 651 - NSA, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Quetol 651 - NSA is a water miscible epoxy resin used for both light and electron microscopy. Its low viscosity permits easy infiltration and embedding, and yields light-colored blocks.

Delivery information: Kit consists of 225 mL Quetol 651, 450 mL NMA , 450 mL NSA, 50 mL DMP-30.

Order Now
 

ORDER

WRITE A REVIEW

REVIEWS (0)

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty