Would you like to log in again?
Paraformaldehyde-Glutaraldehyde Solution Karnovsky's Fixative, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: out of 5
Sort all by:
15720 15730
100503-920EA 284.37 USD InStock
100503-920 100503-482
Paraformaldehyde-Glutaraldehyde Solution Karnovsky's Fixative, Electron Microscopy Science
Stains Microscopy and staining reagents
Paraformaldehyde-glutaraldehyde solution (karnovsky's fixative) is ready to mix.
Paraformaldehyde-Glutaraldehyde Solution is ready to mix. Kit consists of 2 x 10ml ampoules 16% Formaldehyde, 1 x 10ml ampoules 50% Glutaraldehyde, 1 x 50ml 0.2M Sodium Phosphate Buffer
Delivery information: Kit consists of 16% formaldehyde (2×10 ml) ampoules, 50% glutaraldehyde (1×10 ml) ampoules and sodium phosphate buffer 0.2 M, pH 7.2 (1×50 ml).
Order Now
ORDER
SPECIFICATIONS
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty