Would you like to log in again?
Agar, Powder, Bacteriological Grade, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
10200 10202
100505-012EA 53.94 USD InStock
100505-012 100503-640
Agar, Powder, Bacteriological Grade, Electron Microscopy Science
Media for Microbiology Dehydrated Media for Microbiology
Agar, Powder has a gelling temperature (1.5% in H2O) ~35 °C
Gel Strength (1.5% gel) 300g/cm2.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty