Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Aniline blue, certified (C.I. 42755, C.N. DcK-11)

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Acid Blue 22Marine Blue VDisodium hydrogen aminomethyl[[4-[(sulphonatophenyl)amino]phenyl][4-[(sulphonatophenyl)imino]cyclohexa-2,5-dien-1-ylidene]methyl]benzenesulphonateChina BlueCotton BlueCI 42755Blackley BlueCN DcK-11Water Blue

10840
100503-664EA 48.3 USD InStock
100503-664
Aniline blue, certified (C.I. 42755, C.N. DcK-11)
Aniline blue
Used in connective tissue staining procedures to stain collagen. Used in differential staining (Snodgress el al., Lab Invest. 26:329, 1972).

Can be used with biebrich scarlet for staining collagen, reticulum, muscle, plasma, and nuclei. All connective tissues.

For use in staining cytoplasm, cellulose and determining hydrolysis of fats. Can also be used as a contrast stain for carmine and as a counterstain with red nuclear dye.

Solubility : 50% Water; 0.0% Alcohol; 4.5% Cellosolve; 7.8% Glycol; 0.0% Xylene

Certifications: Certified by the Biological Stain Commission.

Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.


Formula: C₃₂H₂₅N₃Na₂O₉S₃
MW: 737.74 g/mol
MDL Number: MFCD00036130
CAS Number: 28631-66-5
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty