Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Azure B (azure I), certified C.I. 52010

Supplier: Electron Microscopy Sciences
Synonyms: Trimethylthionine chlorideC.I. 52010N,N,N′-Trimethylthionin3-(Dimethylamino)-7-(methylamino)phenothiazin-5-ium chlorideMethylene Azur B

11200
100503-670EA 99.48 USD InStock
100503-670
Azure B (azure I), certified C.I. 52010
Azure B (Azure I)
Azur B (Azur I; Methylene Azur B) is an important component of Giemsa's stain for blood protozoa.

Can be used for staining semi-thin sections of plant tissue and distinction of cellular RNA and DNA in botanical tissue. Can also be used as polychromatic stains for thin sections, Negri body stain and malarial parasites.
Hoefert, L.L. (1968). Polychromatic stains for thin sections of Beta embedded in epoxy resin. Stain Tech. 43:145

Solubility : Water-soluble; Alcohol-slightly soluble

Certifications: Certified by the Biological Stain Commission.

Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.


Formula: C₁₅H₁₆ClN₃S
MW: 305.83 g/mol
MDL Number: MFCD00011935
CAS Number: 531-55-5
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/80/28984780.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty