Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Scheduled maintenance of the site is planned from June 8th at 7:00 PM to June 9th at 5:00 PM ET. During this time, contract pricing and registration functionality will be unavailable. We appreciate your patience.

Azure A, certified used in nuclear staining C.I. 52005

Supplier: Electron Microscopy Sciences
Warning
Synonyms: N,N-Dimethylthionine3-Amino-7-(dimethylamino)phenothiazin-5-ium chlorideMethylene Azur A

11240
100503-676EA 134.53 USD InStock
100503-676
Azure A, certified used in nuclear staining C.I. 52005
Azure A
Can be used in nuclear staining; A polychromatic blood stain.

Solubility : Water-soluble; Alcohol-soluble

Certifications: Certified by the Biological Stain Commission.

Caution: All Dyes/Stains are for research use only. Not meant for drug, food household use.


Formula: C₁₄H₁₄ClN₃S
MW: 291.797 g/mol
MDL Number: MFCD00012112
CAS Number: 531-53-3
https://us-legacy.vwr.com/stibo/web/std.lang.all/48/56/28984856.jpg
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty