Would you like to log in again?
Trump's Fixative, Electron Microscopy Science
Supplier: Electron Microscopy Sciences
11750 11752
100503-724EA 94.25 USD InStock
100503-724 100503-726
Trump's Fixative, Electron Microscopy Science
Stains Microscopy and staining reagents
A combination of Sodium Cacodylate, Formalin, and Glutaraldehyde. Ready to use.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty