Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Chloroform ≥99.7%, Glass Distilled

Supplier: Electron Microscopy Sciences
Danger
Synonyms: Methane, trichloro-TrichloroformTrichloromethane

12551 12550
100503-760EA 28.34 USD InStock
100503-760 100503-762
Chloroform ≥99.7%, Glass Distilled
Chloroform
Glass distilled chloroform is available for your convenience.


Formula: CHCl₃
MW: 119.38 g/mol
Boiling Pt: 62 °C (144 °F)
Melting Pt: –63 °C (–81 °F)
Density: 1.47988 g/cm³ (12.35 lbs/gal.)
Flash Pt: 0 °C (32 °F)
Storage Temperature: Ambient
MDL Number: MFCD00000826
CAS Number: 67-66-3
UN: 1888
ADR: 6.1,III
https://us-legacy.vwr.com/stibo/web/std.lang.all/47/98/28984798.jpg
Order Now
Specification Test Results
Assay 99.7% min
Evaporation Residue 3 ppm max.
Water 0.03% max.
pH range 4.6 - 6.2
UV Absorption 1 cm cell vs H₂O 290 nm: 0.01, 270 nm: 0.02, 260 nm: 0.05, 255 nm: 0.15, 245 nm: 1.0

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty