Would you like to log in again?
Graphite Adhesives, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
12660 12650 12661
100503-778EA 12.81 USD InStock
100503-778 100503-776 100503-774
Graphite Adhesives, Electron Microscopy Sciences
Stains Microscopy and staining reagents
Water based and isopropal based graphite adhesives.
  • Water based
  • Isopropal based
  • Graphite extender available
Water based has a flat surface texture. The average flake size is 1 micron. Service temperature is 306°C. Isopropal based has a flat surface texture. The average flake size is 1 micron. Service temperature is 204°C.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty