Would you like to log in again?
Durcupan Water Soluble, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14021 14020 14022
100503-848EA 341.93 USD InStock
100503-848 100503-432 100503-850
Durcupan Water Soluble, Electron Microscopy Sciences
Stains Microscopy and staining reagents
A water soluble embedding medium for EM based on an aliphatic polyepoxide.
Fluka Durcupan embedding agent opens new roads in the observation of enzymatic digestion processes and for carrying out histochemical investigations in the submicroscopic range. It enables formalin and other aqueous fixatives to be used for electron microscopy and the dehydration of the tissue in alcohol or acetone to be dispensed with, so that excessive changes in the ultrastructure of the cells are obviated.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty