Would you like to log in again?
Durcupan ACM, Epoxy Resin, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
14042 14041 14040
100503-854EA 125.34 USD InStock
100503-854 100503-852 100503-434
Durcupan ACM, Epoxy Resin, Electron Microscopy Sciences
Stains Microscopy and staining reagents
An aromatic polyepoxide; a colorless resin of relatively low viscosity, with virtually no shrinkage.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty