Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Eosin Y (yellowish), certified (C.I. 45380)

Supplier: Electron Microscopy Sciences
Warning
Synonyms: Tetrabromofluorescein disodium saltEosin Y disodium saltEosin 2252′,4′,5′,7′-Tetrabromofluorescein disodium saltSolvent Red 43Disodium 2-(2,4,5,7-tetrabromo-6-oxido-3-oxoxanthen-9-yl)benzoateDisodium 2',4',5',7'-tetrabromo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate)Field's Stain B2,4,5,7-Tetrabromofluorescein disodium salt

14850 14851
100503-874EA 60.77 USD InStock
100503-874 100503-872
Eosin Y (yellowish), certified (C.I. 45380)
Eosin Y (yellowish)
Eosin Y can be used with hematoxylin for showing cell nuclei in embryos and kidneys.

  • Component of Wright's stain for blood corpuscles
  • For staining bone marrow to show cell morpholog

Solubility : 44.0% Water; 2.0% Alcohol; 25.0% Cellosolve; 27.5% Glycol; 0.0% Xylene


Formula: C₂₀H₆Br₄Na₂O₅
MW: 691.86 g/mol
Density: 1.018 g/cm³ (8.495 lbs/gal.)
Storage Temperature: Ambient
MDL Number: MFCD00005040
CAS Number: 17372-87-1
UN: 2810
ADR: 6.1,III
Order Now

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty